Dansyl chloride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 5-(Dimethylamino)naphthalene-1-sulphonyl chloride

M-Clarity™ quality level = MQ200

39220-1G-F 39220-50G-F 39220-5G-F
SIAL39220-1G-FEA 127 EUR
SIAL39220-1G-F SIAL39220-50G-F SIAL39220-5G-F
Dansyl chloride, Sigma-Aldrich®
Dansyl chloride
Formula: C₁₂H₁₂ClNO₂S
MW: 269.75 g/mol
Melting Pt: 69…73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003985
CAS Number: 605-65-2
EINECS: 210-092-6
UN: 3261
ADR: 8,III
Merck Index: 13,02840

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR