4-Methoxy-1-naphthol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Methoxynaphthalen-1-ol, 1-Hydroxy-4-methoxynaphthalene
M-Clarity™ quality level = MQ200
174556-25G 174556-5G
SIAL174556-25GEA 325 EUR
SIAL174556-25G SIAL174556-5G
4-Methoxy-1-naphthol, Sigma-Aldrich®
4-Methoxy-1-naphthol
Formula: C₁₁H₁₀O₂
MW: 174.2 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 131 °C
MDL Number: MFCD00003976
CAS Number: 84-85-5
EINECS: 201-566-3