1,8-Naphthalenedicarboxylic anhydride, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Naphthalene-1,8-dicarboxylic anhydride, Benzo[de]isochromene-1,3-dione, 1,8-Naphthalic anhydride
M-Clarity™ quality level = MQ200
N1607-100G N1607-500G
SIALN1607-100GEA 39.6 EUR
SIALN1607-100G SIALN1607-500G
1,8-Naphthalenedicarboxylic anhydride, Sigma-Aldrich®
1,8-Naphthalenedicarboxylic anhydride
Formula: C₁₂H₆O₃
MW: 198.18 g/mol
Boiling Pt: 425
Melting Pt: 267…270 °C
MDL Number: MFCD00006925
CAS Number: 81-84-5
EINECS: 201-380-2