1-Naphthol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1-Hydroxynaphthalene, α-Naphthol, α-Hydroxynaphthalene
M-Clarity™ quality level = MQ200
N2780-100G N2780-10G
SIALN2780-100GEA 822 EUR
SIALN2780-100G SIALN2780-10G
1-Naphthol, Sigma-Aldrich®
1-Naphthol
Formula: C₁₀H₈O
MW: 144.17 g/mol
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 94 °C
Density: 1.224 g/cm³ (20 °C)
Flash Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD00003930
CAS Number: 90-15-3
EINECS: 201-969-4
UN: 2811
ADR: 6.1,III
Merck Index: 13,06409
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b3ec90dc9c3aaa5f4d795a855d487dabe432747&vid=155c70585a9ec4dd427b1a53f47c6c6f137f7055&prid=web&clid=SAPDAM