Silver hexafluoroantimonate 98%
Supplier: Thermo Scientific
Danger
Synonyms: Silver hexafluoroantimonate(V)
312500050 312500250
ACRO312500050EA 132 EUR
ACRO312500050 ACRO312500250
Silver hexafluoroantimonate 98%
Silver hexafluoroantimonate
Formula: AgF₆Sb
MW: 343.62 g/mol
Density: 4,65 to 4.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003401
CAS Number: 26042-64-8
EINECS: 247-429-1
UN: 3260
ADR: 8,II
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Titration Argentometric ≥97.5 %