Molybdenum pentachloride, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

642452-10G 642452-2G
SIAL642452-10GEA 311 EUR
SIAL642452-10G SIAL642452-2G
Molybdenum pentachloride, Sigma-Aldrich®
Molybdenum pentachloride
Formula: Cl₅Mo
MW: 273.2 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 194 °C
Density: 2.928 g/cm³ (20 °C)
MDL Number: MFCD00003467
CAS Number: 10241-05-1
EINECS: 233-575-3
UN: 2508
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR