Chromium(III) oxide, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Chromic oxide
M-Clarity™ quality level = MQ100
634239-100G
634239-25G
SIAL634239-100GEA
594
EUR
SIAL634239-100G
SIAL634239-25G
Chromium(III) oxide, Sigma-Aldrich®
Chromium (III) oxide
Formula:
Cr₂O₃ MW: 151.99 g/mol Boiling Pt: 4000 °C (1013 hPa) Melting Pt: 2435 °C Density: 5.22 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010949 CAS Number: 1308-38-9 EINECS: 215-160-9 Merck Index: 13,02247 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...