Molybdenum(VI) oxide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Molybdenum trioxide
M-Clarity™ quality level = MQ100
203815-25G 203815-5G
SIAL203815-25GEA 291 EUR
SIAL203815-25G SIAL203815-5G
Molybdenum(VI) oxide, Sigma-Aldrich®
Molybdenum (VI) oxide
Formula: MoO₃
MW: 143.94 g/mol
Boiling Pt: 1155 °C (1013 hPa)
Melting Pt: 795 °C
Density: 4.69 g/cm³ (20 °C)
MDL Number: MFCD00003469
CAS Number: 1313-27-5
EINECS: 215-204-7
UN: 3288
ADR: 6.1,III
Merck Index: 13,06261