Cobalt(II) acetylacetonate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Cobalt(II) bis(acetylacetonate), Cobalt (II) bis(2,4-pentanedionato-O,O'), Cobalt (II) 2,4-pentanedionate, Bis(2,4-pentanedionato-O,O')cobalt (II)

M-Clarity™ quality level = MQ200

227129-250G 227129-50G
SIAL227129-250GEA 348 EUR
SIAL227129-250G SIAL227129-50G
Cobalt(II) acetylacetonate, Sigma-Aldrich®
Cobalt (II) acetylacetonate
Formula: C₁₀H₁₄CoO₄
MW: 257.15 g/mol
Melting Pt: 168…171 °C
Density: 0.83 g/cm³ (20 °C)
MDL Number: MFCD00000014
CAS Number: 14024-48-7
EINECS: 237-855-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR