Iron(III) acetylacetonate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Tris(2,4-pentanedionato-O,O')iron (III), Ferric acetylacetonate, Iron (III) 2,4-pentanedionate, Iron (III) tris(2,4-pentanedionato-O,O')
M-Clarity™ quality level = MQ100
F300-100G F300-25G F300-500G
SIALF300-100GEA 62.6 EUR
SIALF300-100G SIALF300-25G SIALF300-500G
Iron(III) acetylacetonate, Sigma-Aldrich®
Iron (III) acetylacetonate
Formula: C₁₅H₂₁FeO₆
MW: 353.18 g/mol
Boiling Pt: 100 °C (9.7513 mmHg)
Melting Pt: 180…183 °C
Density: 5.24 g/cm³ (20 °C)
MDL Number: MFCD00000020
CAS Number: 14024-18-1
EINECS: 237-853-5