Maltol
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-Hydroxy-2-methyl-γ-pyrone, Maltol, 3-Hydroxy-2-methyl-4-pyranone, 3-Hydroxy-2-methyl-4H-pyran-4-one, 3-Hydroxy-2-methyl-4-pyrone
SB00897.1
SB00897.10
SB00897.50
MAYBSB00897.1EA
0
EUR
MAYBSB00897.1
MAYBSB00897.10
MAYBSB00897.50
Maltol
Maltol
Formula:
C₆H₆O₃ MW: 126.11 g/mol Boiling Pt: 285 Density: 1.453 g/cm³ (20 °C) |
MDL Number:
MFCD00006578 CAS Number: 118-71-8 EINECS: 204-271-8 Merck Index: 13,05735 |