Maltol, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
3-Hydroxy-2-methyl-γ-pyrone, Maltol, 3-Hydroxy-2-methyl-4-pyranone, 3-Hydroxy-2-methyl-4H-pyran-4-one, 3-Hydroxy-2-methyl-4-pyrone
M-Clarity™ quality level = MQ100
W265624-1KG-K
W265624-5KG-K
SIALW265624-1KG-KEA
193
EUR
SIALW265624-1KG-K
SIALW265624-5KG-K
Maltol, Sigma-Aldrich®
Maltol
Formula:
C₆H₆O₃ MW: 126.11 g/mol Boiling Pt: 285 °C (1013 hPa) Density: 1.453 g/cm³ (20 °C) |
MDL Number:
MFCD00006578 CAS Number: 118-71-8 EINECS: 204-271-8 Merck Index: 13,05735 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...