Dieldrin
Supplier: EHRENSTORFER GMBH
EHERC12590000EA
68.2
EUR
EHERC12590000
Dieldrin
Dieldrin
Formula:
C₁₂H₈Cl₆O MW: 380,91 g/mol Boiling Pt: 385 °C (1013 hPa) Melting Pt: 174…175 °C Density: 1,75 g/cm³ (20 °C) |
UN:
2761 ADR: 6.1,II Merck Index: 12,03152 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...