Dieldrin

Supplier: LGC Standards PROMOCHEM

PROONMIAP1747EA 253 EUR
PROONMIAP1747
Dieldrin
Dieldrin
Formula: C₁₂H₈Cl₆O
MW: 380,91 g/mol
Boiling Pt: 385 °C (1013 hPa)
Melting Pt: 174…175 °C
Density: 1,75 g/cm³ (20 °C)
UN: 2761
ADR: 6.1,II
Merck Index: 12,03152

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR