1,3-Propanesultone, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1,2-Oxathiolane 2,2-dioxide
M-Clarity™ quality level = MQ200
P50706-100G P50706-1KG P50706-25G
SIALP50706-100GEA 141 EUR
SIALP50706-100G SIALP50706-1KG SIALP50706-25G
1,3-Propanesultone, Sigma-Aldrich®
1,3-Propanesultone
Formula: C₃H₆O₃S
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 31 °C
Density: 1.3996 g/cm³ (35 °C)
MDL Number: MFCD00005355
CAS Number: 1120-71-4
EINECS: 214-317-9
UN: 2811
ADR: 6.1,III