Pyrazine-2-carboxylic acid

Supplier: Thermo Scientific

Synonyms: Pyrazinecarboxylic acid, 2-Pyrazinecarboxylic Acid

CC03401.250 CC03401.1
MAYBCC03401.250EA 0 EUR
MAYBCC03401.250 MAYBCC03401.1
Pyrazine-2-carboxylic acid
Pyrazine-2-carboxylic acid
Formula: C₅H₄N₂O₂
MW: 124.1 g/mol
Boiling Pt: 313 °C (1013 hPa)
MDL Number: MFCD00006130
CAS Number: 98-97-5
EINECS: 202-718-1
Merck Index: 12,08143

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR