2-Isobutyl-3-methoxypyrazine ≥98%
Supplier: Thermo Fisher Scientific
Warning
A13680.03 A13680.06
A13680.03EA 48 EUR
A13680.03 A13680.06
2-Isobutyl-3-methoxypyrazine ≥98%
2-Isobutyl-3-methoxypyrazine
Purity: 98% CAS No.: 24683-00-9 Molecular Formula: C9H14N2O Formula Weight: 166.22 Flash Point: 80°(176°F) Refractive Index: 1.49 MDL No.: MFCD00006128
Formula: C₉H₁₄N₂O
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 107 °C
Density: 0.986
Flash Pt: 80 °C (176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006128
CAS Number: 24683-00-9
EINECS: 246-402-1