2-Acetylpyrazine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Pyrazin-2-ylethan-1-one, Acetylpyrazine
M-Clarity™ quality level = MQ100
251801-1G
251801-5G
SIAL251801-1GEA
27.2
EUR
SIAL251801-1G
SIAL251801-5G
2-Acetylpyrazine, Sigma-Aldrich®
2-Acetylpyrazine
Formula:
C₆H₆N₂O Boiling Pt: 59 °C (33 mmHg) Melting Pt: 79…80 °C |
MDL Number:
MFCD00006134 CAS Number: 22047-25-2 EINECS: 244-753-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...