2-Chloro-4-nitropyridine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
557390-1G
557390-5G
SIAL557390-1GEA
26.1
EUR
SIAL557390-1G
SIAL557390-5G
2-Chloro-4-nitropyridine, Sigma-Aldrich®
2-Chloro-4-nitropyridine
Formula:
C₅H₃ClN₂O₂ MW: 158.54 g/mol Melting Pt: 53.5…54 °C |
MDL Number:
MFCD00661454 CAS Number: 23056-36-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...