4-Hydroxy-3-nitropyridine
Supplier: Thermo Scientific
Synonyms:
3-Nitro-4-pyridon, 3-Nitro-4-pyridone, 3-Nitropyridin-4-ol, 3-Nitro-4-pyridinol
BTB09680.250
BTB09680.1
MAYBBTB09680.250EA
0
EUR
MAYBBTB09680.250
MAYBBTB09680.1
4-Hydroxy-3-nitropyridine
4-Hydroxy-3-nitropyridine
Formula:
C₅H₄N₂O₃ MW: 140.10 g/mol |
MDL Number:
MFCD00128874 CAS Number: 5435-54-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...