2,4,6-Trichloro-1,3,5-triazine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Cyanuric chloride, 2,4,6-Trichloro-s-triazine
M-Clarity™ quality level = MQ200
C95501-1KG
C95501-250G
C95501-5G
SIALC95501-1KGEA
139
EUR
SIALC95501-1KG
SIALC95501-250G
SIALC95501-5G
2,4,6-Trichloro-1,3,5-triazine, Sigma-Aldrich®
2,4,6-Trichloro-1,3,5-triazine
Formula:
C₃Cl₃N₃ MW: 184.41 g/mol Boiling Pt: 190 °C (1013 hPa) Density: 1.32 g/cm³ (20 °C) |
MDL Number:
MFCD00006046 CAS Number: 108-77-0 EINECS: 203-614-9 UN: 2670 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...