Adamantane-1-carboxylic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
106399-100G
106399-25G
SIAL106399-100GEA
101
EUR
SIAL106399-100G
SIAL106399-25G
Adamantane-1-carboxylic acid, Sigma-Aldrich®
Adamantane-1-carboxylic acid
Formula:
C₁₁H₁₆O₂ MW: 180.25 g/mol Melting Pt: 172…175 °C Density: 1.221 g/cm³ (20 °C) |
MDL Number:
MFCD00074720 CAS Number: 828-51-3 EINECS: 212-584-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...