1-Adamantanol
Supplier: Thermo Scientific
Synonyms: 1-Tricyclo[3.3.1.1(3,7)]decanol, 1-Hydroxytricyclo[3.3.1.1(3,7)]decane, Tricyclo[3.3.1.1(3,7)]decan-1-ol, Adamantan-1-ol
SB01577.1 SB01577.10 SB01577.50
MAYBSB01577.1EA 0 EUR
MAYBSB01577.1 MAYBSB01577.10 MAYBSB01577.50
1-Adamantanol
1-Adamantanol
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 85…87 °C
Density: 1.06 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00074729
CAS Number: 768-95-6
EINECS: 212-202-8