2-Adamantanol
Supplier: Thermo Scientific
Synonyms: 2-Hydroxytricyclo[3.3.1.1(3,7)]decane, Tricyclo[3.3.1.1(3,7)]decan-2-ol, 2-Tricyclo[3.3.1.1(3,7)]decanol
SB01544.1 SB01544.10 SB01544.50
MAYBSB01544.1EA 0 EUR
MAYBSB01544.1 MAYBSB01544.10 MAYBSB01544.50
2-Adamantanol
2-Adamantanol
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Melting Pt: 299…303 °C
Density: 1.07 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00074744
CAS Number: 700-57-2
EINECS: 211-846-7