Avantor® Hichrom HI-SE54, GC Columns
Supplier: Avantor
HI05-05-005-2EA 514 EUR
HI05-05-005-2 HI05-05-005-2.5 HI05-05-005-5 HI05-10-010-10 HI05-10-010-15 HI05-10-010-5 HI05-10-020-10 HI05-10-020-15 HI05-10-020-5 HI05-10-040-10 HI05-10-040-15 HI05-10-040-5 HI05-15-015-10 HI05-15-015-15 HI05-15-015-20 HI05-15-015-30 HI05-15-015-5 HI05-25-025-15G5 HI05-25-025-30G5 HI05-25-025-60G5 HI05-25-100-30G5 HI05-32-010-10G2 HI05-32-010-8G2 HI05-32-100-30G5 HI05-53-050-30G10 HI05-53-100-30G5 HI05-18-018-10 HI05-18-018-20 HI05-18-018-40 HI05-18-018-50 HI05-18-018-60 HI05-18-020-10 HI05-18-020-20 HI05-18-020-40 HI05-18-020-50 HI05-18-020-60 HI05-18-040-10 HI05-18-040-20 HI05-18-040-40 HI05-18-040-50 HI05-18-040-60 HI05-20-010-10 HI05-20-010-20 HI05-20-010-40 HI05-20-010-50 HI05-20-010-60 HI05-20-020-10 HI05-20-020-20 HI05-20-020-40 HI05-20-020-50 HI05-20-020-60 HI05-20-040-10 HI05-20-040-20 HI05-20-040-40 HI05-20-040-50 HI05-20-040-60 HI05-20-100-10 HI05-20-100-20 HI05-20-100-40 HI05-20-100-50 HI05-20-100-60 HI05-25-010-15 HI05-25-010-30 HI05-25-010-50 HI05-25-010-60 HI05-25-025-10 HI05-25-025-105 HI05-25-025-15 HI05-25-025-30 HI05-25-025-50 HI05-25-025-60 HI05-25-050-10 HI05-25-050-105 HI05-25-050-15 HI05-25-050-30 HI05-25-050-50 HI05-25-050-60 HI05-25-100-10 HI05-25-100-105 HI05-25-100-15 HI05-25-100-30 HI05-25-100-50 HI05-25-100-60 HI05-32-010-10 HI05-32-010-105 HI05-32-010-15 HI05-32-010-30 HI05-32-010-50 HI05-32-010-60 HI05-32-025-10 HI05-32-025-105 HI05-32-025-15 HI05-32-025-30 HI05-32-025-50 HI05-32-025-60 HI05-32-050-10 HI05-32-050-105 HI05-32-050-15 HI05-32-050-30 HI05-32-050-50 HI05-32-050-60 HI05-32-100-10 HI05-32-100-105 HI05-32-100-15 HI05-32-100-30 HI05-32-100-50 HI05-32-100-60 HI05-32-150-10 HI05-32-150-105 HI05-32-150-15 HI05-32-150-30 HI05-32-150-50 HI05-32-150-60 HI05-32-300-10 HI05-32-300-105 HI05-32-300-15 HI05-32-300-30 HI05-32-300-50 HI05-32-300-60 HI05-32-400-30 HI05-32-500-10 HI05-32-500-105 HI05-32-500-15 HI05-32-500-30 HI05-32-500-50 HI05-32-500-60 HI05-53-010-10 HI05-53-010-15 HI05-53-010-30 HI05-53-025-15 HI05-53-025-30 HI05-53-025-50 HI05-53-025-60 HI05-53-050-10 HI05-53-050-15 HI05-53-050-30 HI05-53-050-50 HI05-53-050-60 HI05-53-100-10 HI05-53-100-15 HI05-53-100-30 HI05-53-100-50 HI05-53-100-60 HI05-53-100-75 HI05-53-150-10 HI05-53-150-15 HI05-53-150-30 HI05-53-150-50 HI05-53-150-60 HI05-53-150-75 HI05-53-265-10 HI05-53-300-10 HI05-53-300-15 HI05-53-300-30 HI05-53-300-50 HI05-53-300-60 HI05-53-300-75 HI05-53-500-10 HI05-53-500-15 HI05-53-500-30 HI05-53-500-50 HI05-53-500-60 HI05-53-500-75 HI05-53-700-15 HI05-53-700-30 HIGCHI05-32-100-25
Avantor® Hichrom HI-SE54, GC Columns
Chromatography Columns
General purpose column for solvent impurities, PCBs, hydrocarbons, essential oils, semivolatiles, allergens, pesticides, etc.
Max temperature limits may change depending on the film thickness.