Silicone fluids, previously Dow Corning 200®
Supplier: DOW CORNING
630068C 630078E 630098Y 630185G
630068CEA 0 EUR
630068C 630078E 630098Y 630185G
Silicone fluids, previously Dow Corning 200®
Circulator Bath Fluids
A range of silicone fluids recommended for use in baths. Low in toxicity, inert and long lasting due to their low volatility.
  • Non corrosive
  • High flash point
  • Long service life
  • Low evaporation
Fluids possess excellent heat transfer characteristics and cover a large temperature range from −40 to +200 °C.
1 cS = 1 mm²/s
Upozornení: Harmful vapours may be given off at elevated temperatures. It is advisable to use this material in a fume cupboard at temperatures greater than 140 °C.