Chemical resistant gloves, A643 Amber Cut
Supplier: Portwest
A643AMRXL A643AMRL A643AMRM
PRTWA643AMRXLEA 79.8 EUR
PRTWA643AMRXL PRTWA643AMRL PRTWA643AMRM
Chemical resistant gloves, A643 Amber Cut
Gloves
A level B cut resistant liner for low risk cut protection and nitrile foam dipping for enhanced grip. Bright orange liner provides easy identification.
  • Material: polyester, elastane
  • ANSI cut level A2
  • 13 gauge liner for a perfect fit
  • 100% breathable seamless liner
  • Palm dipped to increase dexterity and ventilation
  • Nitrile foam coating for excellent grip in wet and dry conditions
Suited for food processing plants and resistant to greases, animal fats and oils.
Certifikace: Standards: AS/NZS 2161 .3, AS/NZS 2161 .2, ANSI/ISEA 105: 2016 CUT Level A2, EN 388:2016 +A1:2018 (4X42B)