ESD safety sandals, Velcro® fastening, Steelite™ Ebro S1 Class 3
Supplier: Portwest
FW48WHR40 FW48WHR41 FW48WHR42 FW48WHR43 FW48WHR44 FW48WHR45 FW48WHR46 FW48WHR36 FW48WHR37 FW48WHR38 FW48WHR39 FW48WHR47 FW48WHR48 FW48WHR34 FW48WHR35
PRTWFW48WHR40EA 53.4 EUR
PRTWFW48WHR40 PRTWFW48WHR41 PRTWFW48WHR42 PRTWFW48WHR43 PRTWFW48WHR44 PRTWFW48WHR45 PRTWFW48WHR46 PRTWFW48WHR36 PRTWFW48WHR37 PRTWFW48WHR38 PRTWFW48WHR39 PRTWFW48WHR47 PRTWFW48WHR48 PRTWFW48WHR34 PRTWFW48WHR35
ESD safety sandals, Velcro® fastening, Steelite™ Ebro S1 Class 3
Shoes and Boots
ESD safety sandal with perforated upper that offers supreme breathability. Hook and loop fastening feature ensures optimum fit and convenient access for the wearer.
  • CE certified
  • 200 joule steel toecap
  • Anti-static footwear
  • Energy absorbing seat region
  • Slip resistant outsole (SRC)
  • Oil resistant outsole
Superb comfort and fit with steel safety components. The enhanced outsole properties for excellent grounding of static build up that may damage sensitive electronic components.
Innovation, quality components, styling and expert construction define the Portwest® footwear collections. Benefits include Machine washable at 30 °C, breathable to keep your feet comfortable, ideal for the medical and food Industries, allows rapid dawning.
Certifikace: EN 61340-4-3 Class 3
EN ISO 20345:2011