Anti impact gloves, Aqua-Seal Pro A726
Supplier: Portwest
A726O4RL A726O4RXL
PRTWA726O4RLEA 36.7 EUR
PRTWA726O4RL PRTWA726O4RXL
Anti impact gloves, Aqua-Seal Pro A726
Gloves
The Aqua-Seal Pro is 100% waterproof and offers comprehensive protection for the hand against multiple hazards. TPR protective pods across the back of the hand provides protection against impact risks to the hand. A 100 gram Insulated lining gives excellent thermal insulation properties.
  • TPR pod coverage
  • Reflective trimmings
  • PVC dotted palm for enhanced grip
  • Highly durable synthetic leather with elastane and neoprene with 0,0936 m³ surface area
  • Low linting construction for minimal contamination
This range of gloves provides vital protection against impact to the hand from multiple hazards.
Certifikace: AS/NZS 2161 .2; AS/NZS 2161 .3; AS/NZS 2161 .5; EN 388:2003 (4332); EN 388:2016 (3233XX); ANSI/ISEA 105 - 2011 Abrasion Level (3). EN 511 (221)