High visibility rain jackets, H440
Supplier: Portwest
H440YERL H440YERM H440YERXL H440YER4XL H440YERXXL H440YERXXXL H440YERS H440YERXS H440ORR4XL H440ORR5XL H440ORRL H440ORRM H440ORRS H440ORRXL H440ORRXS H440ORRXXL H440ORRXXXL
PRTWH440YERLEA 33.4 EUR
PRTWH440YERL PRTWH440YERM PRTWH440YERXL PRTWH440YER4XL PRTWH440YERXXL PRTWH440YERXXXL PRTWH440YERS PRTWH440YERXS PRTWH440ORR4XL PRTWH440ORR5XL PRTWH440ORRL PRTWH440ORRM PRTWH440ORRS PRTWH440ORRXL PRTWH440ORRXS PRTWH440ORRXXL PRTWH440ORRXXXL
High visibility rain jackets, H440
Workwear
Designed to keep the wearer visible, safe and dry in foul weather conditions, the H440 is extremely practical and waterproof. This garment can be easily rolled up and stored when not in use.
  • Reflective tape for increased visibility
  • Two pockets for secure storage
  • Deep storage pockets
  • Pack away hood for added functionality
  • Inner elasticated cuff
  • 50+ UPF rated fabric to block 98% of UV rays
Shell fabric: 100% polyester, 190T, plain weave, PVC coated 200 g
Lining fabric: Yoke only: 100% polyester mesh 60 g
Features include adjustable cord and toggle for a secure fit, vented back yoke and eyelets for added breathability and comfort. Extremely water resistant fabric finish, water beads away from fabric surface.
Certifikace: ANSI/ISEA 107: 2015 TYPE R CLASS 3; AS 4602.1 Class D/N; EN 343:2019 Class 3:1 X WP 5,000mm; EN ISO 20471 Class 3