All weather outdoor jackets, Bizflame Rain Hi-Vis, S779
Supplier: Portwest
S779ONRS S779ONRXXL S779ONRXXXL S779ONRM S779ONRL S779ONRXL
PRTWS779ONRSEA 232 EUR
PRTWS779ONRS PRTWS779ONRXXL PRTWS779ONRXXXL PRTWS779ONRM PRTWS779ONRL PRTWS779ONRXL
All weather outdoor jackets, Bizflame Rain Hi-Vis, S779
Workwear
This multi-protection waterproof, breathable jacket offers protection against multiple risks. A detachable FR fleece liner makes this jacket suitable for use in all weather conditions. Jacket to be worn with trousers for full protection.
  • Flame resistant treated waterproof fabric prevents water penetration
  • Premium sew on flame resistant reflective tape
  • FR cotton lined for added warmth and comfort
  • Detachable lining for added versatility in all weathers conditions
  • Five pockets for ample storage
  • Concealed phone pocket
Features include breathable fabric to draw moisture away from the body keeping the wearer cool, dry and comfortable. Taped seams provide additional protection. Two-way zip for quick and easy access.
Shell fabric: Bizflame rain 250 g
Lining fabric: 100% cotton flame resistant lining 170 g
Additional lining fabric: Removable flame resistant fleece liner 350 g
Certifikace: EN ISO 14116 Index 3; EN 1149 -5; RIS-3279-TOM ISSUE 1 (ORANGE ONLY); EN 13034 Type 6; EN ISO 20471 Class 3; EN 343 Class 3:3; CE-CAT III.
Upozornení: Use non-biological powder. Do not use softener.