General purpose gloves, Touchscreen, A195
Supplier: Portwest
A195U8RL A195U8RM A195U8RS A195U8RXL A195U8RXS A195U8RXXL
PRTWA195U8RLEA 20.3 EUR
PRTWA195U8RL PRTWA195U8RM PRTWA195U8RS PRTWA195U8RXL PRTWA195U8RXS PRTWA195U8RXXL
General purpose gloves, Touchscreen, A195
Gloves
Gloves made of PU and designed specifically for touchscreen devices. Perfect for intricate tasks.
  • Seamless 13 gauge liner
  • 100% breathable seamless liner
Ideal for use in electronics assembly, testing and precision work.
Colour: Purple/Black
Certifikace: EN 420, EN 388:2016 +A1:2018 (2.1.2.1.X), ANSI/ISEA 105: 2016 cut Level A1