ESD safety sandals, Compositelite™ ESD Tagus (S1P), FC04
Supplier: Portwest
FC04BKR39 FC04BKR40 FC04BKR43 FC04BKR44 FC04BKR45 FC04BKR46 FC04BKR47 FC04BKR41 FC04BKR42
PRTWFC04BKR39EA 62.6 EUR
PRTWFC04BKR39 PRTWFC04BKR40 PRTWFC04BKR43 PRTWFC04BKR44 PRTWFC04BKR45 PRTWFC04BKR46 PRTWFC04BKR47 PRTWFC04BKR41 PRTWFC04BKR42
ESD safety sandals, Compositelite™ ESD Tagus (S1P), FC04
Shoes and Boots
100% non metallic durable safety sandal with dual density PU outsole, certified to ESD Class 3. Breathable upper design for those working in warm conditions. Hook and loop closing for snug fit.
  • Composite toe cap (200 J)
  • Composite midsole
  • Anti-static footwear
  • Energy absorbing seat region
  • Slip resistant outsole (SRC)
CompositeLite™ ESD quality non metallic safety components and enhanced outsole properties for excellent grounding of static build up that may damage sensitive electronic components.
Split Nubuck leather upper and PU/PU
outsole.
Certifikace: EN 61340-4-3 Class 3, EN ISO 20345:2011.