VWR® Drosophila Vials
Supplier: VWR Collection
734-2265EA 579 EUR
734-2265 734-2254 734-2255 734-2257 734-2258 734-2259 734-2261 734-2262 734-2264
VWR® Drosophila Vials
Vials
For storage, handling and transportation of Drosophila flies.
Manufactured in ISO 9001:2000 certified facility. Narrow and wide vials are manufactured in PP, PS or K-Resin and packed in bulk or tray formats.
Polystyrene (PS): This resin has glass-like clarity, allowing very clear, distortion-free viewing of fly gender and behaviour.
Polypropylene (PP), autoclavable: PP is a clear resin that is softer than PS. Ideal for cost-sensitive experiments and clear viewing.
K-Resin: K-Resin is a premium scratch resistant resin, combining the clarity of PS with the flexibility of PP.
Upozornení: *The product(s) on this page are outside the scope of the EU Directive 2019/904.