VWR® Test Tube Racks, Pivoting Stainless Steel
Supplier: VWR Collection
444-2955EA 325 EUR
444-2955 444-2956 444-2957 444-2958 444-2959
VWR® Test Tube Racks, Pivoting Stainless Steel
Racks Tube Racks
Informace o príslušenství: * Platform supplied with mini shaker, incubating mini shaker, microplate shaker and incubating microplate shaker. Note other shakers require an optional perforated platform to enable flask clamps or test tube racks to be mounted which must be ordered separately.