Racks for cryoboxes, half box racks
Supplier: TENAK
TE21410 TE21414 TE21412 TE21416 TE21418 TE21420 TE21422
479-0856EA 70.6 EUR
479-0856 479-0858 479-0857 479-0859 479-0860 TENATE21420 TENATE21422
Racks for cryoboxes, half box racks
Racks Cryogenic Box Racks
Classic design of stainless steel rack for storage of cryoboxes in liquid nitrogen.
  • Can be used with all brands of chest type freezers
  • Suitable for all small cyroboxes with footprint 80×80 mm and height 50 mm
  • The cryoboxes are secured with a locking rod
  • Suitable for cryoboxes in cardboard, polypropylene and polycarbonate
Informace pro objednávání: Cryoboxes must be ordered separately.