Blotting paper, Amersham™ Hybond™
Supplier: Cytiva
Amersham™ Hybond™
RPN6101MEA 88.54 EUR
RPN6101M
Blotting paper, Amersham™ Hybond™
Blotting Papers
Research laboratory blotting paper for use in gel electrophoresis in all molecular biology and protein studies applications.
  • Recommended for use with all Hybond™ blotting membranes
  • High purity and uniformity ensures consistent and reproducible results
  • Pure Cellulose Grade, 0,39 mm thick paper with medium wet strength and smooth surface