Refrigerated and heating circulators, MAGIO™ MS series
Supplier: JULABO GmbH
9032704 9032705 9032707 9032713.N1 9032713.S1 9032714.N1 9032714.S1 9032716.N1 9032715.N1 9032717.N1 9032728.N1 9032757
461-1429EA 7960 EUR
461-1429 461-1430 461-1432 461-1433 461-1434 462-0638 462-0639 462-0640 JULA9032715.N1 JULA9032717.N1 JULA9032728.N1 JULA9032757
Refrigerated and heating circulators, MAGIO™ MS series
Circulating Thermostats Open Circulator Systems Heating/Cooling Circulators
The MAGIO™ series of cooling and heating circulators operate with a working temperature range of –50 to +300 °C and heating capacity of up to 3 kW. With their powerful pressure/suction pump the MAGIO™ units are ideal for external temperature applications. An extensive range of accessories and excellent dynamics mean that the MAGIO™ circulators can be modularly and individually adapted to applications.
  • Bright, easy to read display
  • Integrated programmer allows temperature time profiles to be run
  • Stainless steel wetted parts mean units are ideal for demanding external applications
  • Automatic shut off in the event of high temperature or low liquid level to avoid any potential damage
  • Bath tanks have high quality thermal insulation to save energy and a built-in drain tap
  • Powerful, continuously adjustable pressure/suction pump
  • Choice of models with environmentally-friendly natural refrigerant
The high resolution, modern colour TFT touch display has a multi-lingual user interface and shows all important information at a glance. Three large, predefined main screens clearly display data and graphics with various application priorities. All display functions are easy to operate with a fingertip.
Modern interfaces make remote control easy and practical data management possible as well as allowing integration into process structures. All MAGIO™ units have integrated PT100 connection, USB interface, RS232 and Ethernet. Analogue interfaces are available as optional accessories.
Max. flow rate/max. pressure: 31 L/min; 0,92 bar (pressure), 0,4 bar (suction)
Certifikace: Classification III according to DIN 12876-1. IP protection class according to DIN EN 60529: IP 21.
Delivery information: Supplied with bath cover and two each barbed fitting for tubing 8 and 12 mm inner Ø (pump connections M16×1 male).
Upozornení: Note: Cooling capacity at +20; 0; −10; −20; −30 and −40 °C