J.T.Baker® BAKERBOND®, Ion Exchange Chromatography Resin, PolyCSX Multimode
Supplier: Avantor
J.T.Baker® BAKERBOND®
7587-04EA 0 EUR
7587-04 7587-05 7587-02 7587-03 7587-01
J.T.Baker® BAKERBOND®, Ion Exchange Chromatography Resin, PolyCSX Multimode
Chromatography Sorbents HPLC Sorbents
The J.T.Baker® BAKERBOND® PolyCSX multimode ion exchange (IEX) chromatography resin functions as a strong cation exchange resin with improved selectivity for superior separation in process chromatography applications.
Powered by a proprietary surface chemistry combining strong cation exchange functionality with weak cationic and anionic exchange sites, BAKERBOND® PolyCSX multimode cation exchange chromatography media utilizes its unique selectivity to enable higher purification performance across a wide range of biopharmaceuticals.
Designed and manufactured by Avantor to the high standards established for our J.T.Baker® brand chemicals, BAKERBOND® PolyCSX ion exchange resin can be used with other J.T.Baker® BAKERBOND® resins to enhance the separation power of your chromatography process steps.
Informace pro objednávání: 1 ml and 5 ml lab columns are also available under complementary products.