Khellin ≥95%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Visammin, 4,9-Dimethoxy-7-methyl-5H-furo[3,2-g]chromen-5-one, 4,9-Dimethoxy-7-methyl-5H-furo[3,2-g][1]benzopyran-5-one
461800050 461800250
ACRO461800050EA 82.4 EUR
ACRO461800050 ACRO461800250
Khellin ≥95%
Khellin
  • Bioactive
Formula: C₁₄H₁₂O₅
MW: 260.25 g/mol
Melting Pt: 150…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00005007
CAS Number: 82-02-0
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Light yellow to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 150 to 154 °C
Water ≤1 %
GC ≥94 %