7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin (CPM), Sigma-Aldrich®
Supplier: SIGMA-ALDRICH MICROSCOPY
C1484-25MGEA 521 EUR
C1484-25MG
7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin (CPM), Sigma-Aldrich®
7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin (CPM)
7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin has been used: as a nontoxic substitute for conjugation with antibody(93) to aid fluorescent detection of Coenzyme A (CoA-SH) in human N-myristoyltransferases assay (94)
Formula: C₂₄H₂₂N₂O₄
MW: 402,44 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00077334
CAS Number: 76877-33-3