DAF-2 diacetate, Sigma-Aldrich®

Supplier: SIGMA-ALDRICH MICROSCOPY

D2813-1MGEA 718 EUR
D2813-1MG
DAF-2 diacetate, Sigma-Aldrich®
DAF-2 diacetate

Cell permeable fluorescent detector of nitric oxide in living cells. 4,5-Diaminofluorescein diacetate (DAF-2 DA) has been used as a fluorescent dye for NO (nitric oxide) detection.

Formula: C₂₄H₁₈N₂O₇
MW: 446,41 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05865101
CAS Number: 205391-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR