Crocein scarlet 7B, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Synonyms: C.I. 27165, Acid Red 71
C8822-1G C8822-25G C8822-5G
C8822-1GEA 42.1 EUR
C8822-1G C8822-25G C8822-5G
Crocein scarlet 7B, Sigma-Aldrich®
Crocein scarlet 7B
Crocein Scarlet 7B, (Acid Red 71)
Formula: C₂₄H₁₈N₄Na₂O₇S₂
MW: 584.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00056588
CAS Number: 6226-76-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5ce92fec0125b755a0f34d5ca3be5b4386180e1&vid=b4717aa3643ea69107eaabcfcbbb282750af453e&prid=web&clid=SAPDAM