Thioflavin S, Sigma-Aldrich®

Supplier: SIGMA-ALDRICH MICROSCOPY
Warning

T1892-25GEA 82.6 EUR
T1892-25G
Thioflavin S, Sigma-Aldrich®
Thioflavin S

Thioflavine S has been used to stain and visualize β-amyloid structures in the tissue sections.

Storage Temperature: Ambient
MDL Number: MFCD00071453
CAS Number: 1326-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR