Molybdenum Blue reagent, spray reagent, Sigma-Aldrich®

Supplier: SIGMA-ALDRICH MICROSCOPY
Danger

M1942-100MLEA 110 EUR
M1942-100ML
Molybdenum Blue reagent, spray reagent, Sigma-Aldrich®
Molybdenum Blue reagent

For use in the detection of phospholipids and related compounds.

Storage Temperature: Ambient
ADR: 8,2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR