Haematoxylin solution (Mayer), Sigma-Aldrich®
Supplier: SIGMA-ALDRICH MICROSCOPY
Warning
MHS16-500MLEA 63.5 EUR
MHS16-500ML
Haematoxylin solution (Mayer), Sigma-Aldrich®
Haematoxylin staining solutions
General purpose nuclear stain, progressive type. Used with hematoxylin and eosin staining.
Formula: C₁₆H₁₄O₆
MW: 302,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078111
CAS Number: 517-28-2
IVDLogo