Oil Red EGN, Sigma-Aldrich®

Supplier: SIGMA-ALDRICH MICROSCOPY
Warning

234117-25GEA 91.9 EUR
234117-25G
Oil Red EGN, Sigma-Aldrich®
Oil Red EGN

Oil Red EGN has been used to stain lipid droplets accumulated in cell during adipogenesis.

Formula: C₂₅H₂₂N₄O
MW: 394,47 g/mol
Melting Pt: 130 °C (dec., lit.)
Storage Temperature: Ambient
MDL Number: MFCD00009688
CAS Number: 4477-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR