N-Benzylidenebenzylamine, Sigma-Aldrich®

Supplier: SIGMA-ALDRICH MICROSCOPY
Warning

231304-5GEA 95.2 EUR
231304-5G
N-Benzylidenebenzylamine, Sigma-Aldrich®
N-Benzylidenebenzylamine

Indicator for organolithium assay.

Formula: C₁₄H₁₃N
MW: 195,26 g/mol
Density: 1,038 g/mL (25 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00010006
CAS Number: 780-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR