1,3-Diiminoisoindoline ≥97%, Sigma-Aldrich®

Supplier: SIGMA-ALDRICH MICROSCOPY
Danger

279528-25GEA 468 EUR
279528-25G
1,3-Diiminoisoindoline ≥97%, Sigma-Aldrich®
1,3-Diiminoisoindoline

1,3-Diiminoisoindoline,97% (1,3-Isoindolinediimine; Phthalimide diimide)

Formula: C₈H₇N₃
MW: 145,17 g/mol
Melting Pt: ~197 °C (dec., lit.)
Storage Temperature: Ambient
MDL Number: MFCD00010062
CAS Number: 57500-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR