Gallocyanine 90% (dye content), Sigma-Aldrich®

Supplier: SIGMA-ALDRICH MICROSCOPY
Warning

124508-10GEA 87.6 EUR
124508-10G
Gallocyanine 90% (dye content), Sigma-Aldrich®
Gallocyanine

Gallocyanine, Dye content 90 % (7-Dimethylamino-4-hydroxy-3-oxo-phenoxazine-1-carboxylic acid)

Formula: C₁₅H₁₃ClN₂O₅
MW: 336,73 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011926
CAS Number: 1562-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR